Copper(I) chloride 99%, extra pure, purified

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cuprous chloride, Copper chloride

AC20839-0250 AC20839-1000 AC20839-5000
TS20839-0250EA 111 USD
TS20839-0250 TS20839-1000 TS20839-5000
Copper(I) chloride 99%, extra pure, purified
Copper (I) chloride
Formula: ClCu
MW: 99 g/mol
Boiling Pt: 1367
Melting Pt: 430
Density: 4.14 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010971
CAS Number: 7758-89-6
EINECS: 231-842-9
UN: 2802
ADR: 8 III
Merck Index: 13,02690

Order Now

Specification Test Results

Appearance Light green to gray Crystalline powder
Titration Complexometric 98.5 to 101.5 %
Sulfate (SO4) ≤500 ppm
Iron (Fe) ≤200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR