(3-Glycidoxypropyl)trimethoxysilane 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (3-Glycidyloxypropyl)trimethoxysilane, 3-Glycidoxypropyltrimethoxysilane, (3-(2,3-Epoxypropoxy)propyl)trimethoxysilane

AC21654-0025 AC21654-0050 AC21654-1000 AC21654-5000
TS21654-0025EA 616 USD
TS21654-0025 TS21654-0050 TS21654-1000 TS21654-5000
(3-Glycidoxypropyl)trimethoxysilane 97%
(3-Glycidoxypropyl)trimethoxysilane
Formula: C₉H₂₀O₅Si
Boiling Pt: 120 °C (2 mmHg)
Density: 1.07 g/cm³
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00005144
CAS Number: 2530-83-8
EINECS: 219-784-2

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4280 to 1.4300 (20°C, 589 nm)
GC ≥96.0 %
Color scale ≤50 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR