Manganese(II) sulfate monohydrate 98+% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Manganese sulphate monohydrate, Manganese sulfate monohydrate, Manganous sulfate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate, Manganese(II) sulfate hydrate

AC42391-0025 AC42391-0050 AC42391-5000
TS42391-0025EA 315 USD
TS42391-0025 TS42391-0050 TS42391-5000
Manganese(II) sulfate monohydrate 98+% ACS
Manganese (II) sulphate monohydrate
Formula: MnSO₄·1H₂O
MW: 169.02 g/mol
Melting Pt: 700
Storage Temperature: Ambient
MDL Number: MFCD00149159
CAS Number: 10034-96-5
EINECS: 232-089-9
UN: 3077
ADR: 9 III
Merck Index: 13,05763

Order Now

Specification Test Results

Appearance Light pink Powder
Titration Complexometric 98.0 to 101.0 %
Chloride (Cl) ≤50 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤20 ppm
Magnesium (Mg) ≤50 ppm
Nickel (Ni) ≤200 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤500 ppm
Zinc (Zn) ≤50 ppm
Heavy metals (as Pb) ≤20 ppm
Insoluble matter ≤0.01 %
KMnO4-reducing substance Passes test
Loss on ignition 10.0 % to 12.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR