Mercury(II) thiocyanate 99+%

Supplier: Thermo Scientific Chemicals
Danger

AC19689-0500 AC19689-2500
TS19689-0500EA 144 USD
TS19689-0500 TS19689-2500
Mercury(II) thiocyanate 99+%
Mercury (II) thiocyanate
Formula: C₂HgN₂S₂
Melting Pt: 165 °C (dec.)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00011039
CAS Number: 592-85-8
EINECS: 209-773-0
UN: 1646
ADR: 6.1,II
Merck Index: 13,05917

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Titration Complexometric ≥99.0 %
Iron (Fe) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR