Ammonium iodide 99%, Extra Pure

Supplier: Thermo Scientific Chemicals
Warning

AC44807-0025 AC44807-1000 AC44807-5000
TS44807-0025EA 805 USD
TS44807-0025 TS44807-1000 TS44807-5000
Ammonium iodide 99%, Extra Pure
Ammonium iodide
Formula: NH₄I
MW: 144.94 g/mol
Melting Pt: 551 °C (subl.)
Density: 2.51 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011424
CAS Number: 12027-06-4

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder (May turn yellow on storage)
Titration Argentometric 98.5 to 101.5 %
Residue after ignition ≤0.05 %
Heavy metals (as Pb) ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR