Cesium iodide 99.999% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC19282-0010
AC19282-0100
AC19282-0500
TS19282-0010EA
35
USD
TS19282-0010
TS19282-0100
TS19282-0500
Cesium iodide 99.999% (trace metals basis)
Cesium iodide
Formula:
CsI Boiling Pt: 1280 °C (760 mmHg) Melting Pt: 626 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010962 CAS Number: 7789-17-5 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | White Crystalline powder |
Total trace metal impurities | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...