Manganese(II) chloride, anhydrous 99+% 80 mesh

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Manganous chloride, Manganese dichloride

AC22361-0100 AC22361-0500
TS22361-0100EA 68 USD
TS22361-0100 TS22361-0500
Manganese(II) chloride, anhydrous 99+% 80 mesh
Manganese (II) chloride
Formula: MnCl₂
MW: 125.84 g/mol
Boiling Pt: 1190
Melting Pt: 650
Storage Temperature: Ambient
MDL Number: MFCD00011114
CAS Number: 7773-01-5
EINECS: 231-869-6
UN: 3077
ADR: 9 III
Merck Index: 12,05768

Order Now

Specification Test Results

Appearance Pink Powder
Titration Complexometric ≥99.0 %
Solubility (10 % in dilute HCl) Clear solution
Particle size -80 mesh


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR