Chromium(VI) oxide 98+% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chromic anhydride, Chromium trioxide

AC40523-0010 AC40523-5000
TS40523-0010EA 1048 USD
TS40523-0010 TS40523-0025 TS40523-5000
Chromium(VI) oxide 98+% ACS
Chromium (VI) oxide
Formula: CrO₃
Melting Pt: 196 °C
Density: 2.7 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010952
CAS Number: 1333-82-0
EINECS: 215-607-8
UN: 1463
ADR: 5.1,II
REACH: 01-2119458868-17
Merck Index: 13,02256

Order Now

Specification Test Results

Appearance Red-brown to purple Powder, crystals or granules
Titration Iodometric ≥98.0 %
Chloride (Cl) ≤50 ppm
Nitrate (NO3) ≤500 ppm
Sulfate (SO4) ≤50 ppm
Aluminium (Al) ≤200 ppm
Barium (Ba) ≤100 ppm
Iron (Fe) ≤200 ppm
Sodium (Na) ≤0.2 %
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR