Phosphorus(V) bromide 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Phosphorus pentabromide
AC42827-0250
AC42827-1000
TS42827-0250EA
94
USD
TS42827-0250
TS42827-1000
Phosphorus(V) bromide 95%
Phosphorus (V) bromide
Formula:
PBr₅ MW: 430.49 g/mol Melting Pt: 105…107 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011437 CAS Number: 7789-69-7 UN: 2691 ADR: 8,II |
Specification Test Results
Appearance | Yellow to orange to red Crystalline powder or crystals and/or chunks |
Titration Argentometric | ≥94 % |
Insoluble matter | Pass |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...