Strontium chloride, anhydrous 99.99% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC36974-0050
AC36974-0250
TS36974-0050EA
106
USD
TS36974-0050
TS36974-0250
Strontium chloride, anhydrous 99.99% (trace metals basis)
Strontium chloride
Formula:
Cl₂Sr MW: 158.53 Boiling Pt: 1250 Melting Pt: 868 Density: 1.96 |
MDL Number:
MFCD00011249 CAS Number: 10476-85-4 EINECS: 233-971-6 Merck Index: 12,09000 |
Specification Test Results
Appearance | White Crystalline powder |
Assay | 54.4 to 55.9 % (Sr) |
Total trace metal impurities | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...