Sodium hexafluorophosphate 98.5+%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC19682-0100
AC19682-0500
AC19682-2500
TS19682-0100EA
54
USD
TS19682-0100
TS19682-0500
TS19682-2500
Sodium hexafluorophosphate 98.5+%, pure
Sodium hexafluorophosphate
Formula:
F₆NaP MW: 167.95 g/mol Melting Pt: 200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011122 CAS Number: 21324-39-0 EINECS: 244-333-1 UN: 3260 ADR: 8,II |
Specification Test Results
Appearance | White to light gray Crystalline powder |
Assay | ≥98.50 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...