Potassium chromate 99.5% for analysis

Supplier: Thermo Scientific Chemicals
Danger

AC20234-0025 AC20234-5000
TS20234-0025EA 446 USD
TS20234-0025 TS20234-5000
Potassium chromate 99.5% for analysis
Potassium chromate
Formula: K₂CrO₄
MW: 194.19 g/mol
Boiling Pt: 1000
Melting Pt: 985
Storage Temperature: Ambient
MDL Number: MFCD00011368
CAS Number: 7789-00-6
EINECS: 232-140-5
UN: 3077
ADR: 9 III
Merck Index: 12,07784

Order Now

Specification Test Results

Appearance Yellow Crystals
Titration Iodometric ≥99.5 %
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤100 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤20 ppm
Sodium (Na) ≤500 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR