Sodium chlorate 99% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC44641-1000
AC44641-5000
TS44641-1000EA
72
USD
TS44641-1000
TS44641-5000
Sodium chlorate 99% ACS
Sodium chlorate
Formula:
ClNaO₃ MW: 106.44 g/mol Boiling Pt: 300 Melting Pt: 260 Density: 2.49 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003479 CAS Number: 7775-09-9 UN: 1495 ADR: 5.1 II Merck Index: 13,08670 |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Titration | ≥99.0 % |
Chloride (Cl) | ≤0.005 % |
Bromate (BrO3) | ≤0.015 % |
Sulfate (SO4) | ≤0.001 % |
Calcium (Ca) | ≤0.002 % |
Iron (Fe) | ≤0.0005 % |
Magnesium (Mg) | ≤0.002 % |
Potassium (K) | ≤0.01 % |
Heavy metals (as Pb) | ≤0.001 % |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...