Sodium chlorate 99+%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

AC22322-0010 AC22322-2500
TS22322-0010EA 73 USD
TS22322-0010 TS22322-2500
Sodium chlorate 99+%, extra pure
Sodium chlorate
Formula: ClNaO₃
MW: 106.44 g/mol
Boiling Pt: 300
Melting Pt: 260
Density: 2.49 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003479
CAS Number: 7775-09-9
EINECS: 231-887-4
UN: 1495
ADR: 5.1 II
REACH: 01-2119474389-23
Merck Index: 13,08670

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Water ≤0.05 %
Titration ≥99.0 %
Sodium chloride (NaCl) ≤1000 ppm
Insoluble matter ≤0.005 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR