Sodium carbonate, anhydrous 99.95%, extra pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Soda, Carbonic acid sodium salt, Calcined soda, Disodium carbonate, Carbonic acid disodium salt, Na-X, Bisodium carbonate, Carbonic acid, disodium salt, Soda ash
AC20776-0025
AC20776-5000
TS20776-0025EA
283
USD
TS20776-0025
TS20776-5000
Sodium carbonate, anhydrous 99.95%, extra pure
Sodium carbonate
Formula:
Na₂CO₃ MW: 105.99 g/mol Boiling Pt: 1600 Melting Pt: 854 Storage Temperature: Ambient |
MDL Number:
MFCD00003494 CAS Number: 497-19-8 EINECS: 207-838-8 REACH: 01-2119485498-19 Merck Index: 14,08596 |
|
Specification Test Results
Appearance | White Crystalline powder |
Loss on drying | ≤1 % (285°C) |
Titration with HCl | ≥99.90 % (on dried substance) |
Chloride (Cl) | ≤20 ppm |
Phosphate (PO4) | ≤10 ppm |
Aluminium (Al) | ≤10 ppm |
Calcium (Ca) | ≤50 ppm |
Iron (Fe) | ≤5 ppm |
Potassium (K) | ≤100 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Total nitrogen compounds | ≤0.001 % (as N) |
Silicon dioxide (SiO2) | ≤50 ppm |
Total sulfur (as SO4) | ≤0.005 % |
Insoluble matter | ≤0.01 % (in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...