Lithium carbonate 98+% for analysis

Supplier: Thermo Scientific Chemicals
Warning

AC44632-0010 AC44632-2500
TS44632-0010EA 204 USD
TS44632-0010 TS44632-2500
Lithium carbonate 98+% for analysis
Lithium carbonate
Formula: Li₂CO₃
MW: 73.89 g/mol
Melting Pt: 720 °C
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2

Order Now

Specification Test Results

Appearance White Crystalline powder
Assay ≥98.0 %
Chloride (Cl) ≤0.005 %
Nitrate (NO3) ≤0.005 %
Sulfate (SO4) ≤0.3 %
Iron (Fe) ≤0.005 %
Sodium (Na) ≤0.1 %
Heavy metals (as Pb) ≤0.005 %
Insoluble matter ≤0.05 % (in HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR