Potassium bromate ACS

Supplier: Thermo Scientific Chemicals
Danger

AC44694-1000 AC44694-5000
TS44694-1000EA 75 USD
TS44694-1000 TS44694-5000
Potassium bromate ACS
Potassium bromate
Formula: KBrO₃
MW: 167 g/mol
Melting Pt: 350 °C
Density: 3.27 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011359
CAS Number: 7758-01-2
UN: 1484
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Iodometric ≥99.8 % (Dried basis)
pH 5.0 to 9.0 (5% soln. at 25°C)
Bromide (Br) Passes test (Limit about 0.05%)
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤0.002 %
Sodium (Na) ≤0.01 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR