Sodium borohydride 99%, powder, VenPure™ SF, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate

AC44848-0025 AC44848-0100 AC44848-1000 AC44848-5000
TS44848-0025EA 816 USD
TS44848-0025 TS44848-0100 TS44848-1000 TS44848-5000
Sodium borohydride 99%, powder, VenPure™ SF, pure
Sodium borohydride
Formula: NaBH₄
MW: 37.83 g/mol
Melting Pt: 400 (dec.)
Flash Pt: 69
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
UN: 1426
ADR: 4.3 I
Merck Index: 13,08664

Order Now

Specification Test Results

Appearance White Fine powder
Lightness 91.4 to 100 (lightness scale)
Titration with Na2S2O3 ≥98.5 %
Silicate (as SiO2) 0.0021 to 0.30 %
Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR