Sodium borohydride 98%, pellets (1-2 cm)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate

AC20148-0025 AC20148-1000 AC20148-5000
TS20148-0025EA 947 USD
TS20148-0025 TS20148-1000 TS20148-5000
Sodium borohydride 98%, pellets (1-2 cm)
Sodium borohydride
Formula: NaBH₄
MW: 37.83 g/mol
Melting Pt: 400 (dec.)
Flash Pt: 69
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3 I
Merck Index: 13,08664

Order Now

Specification Test Results

Appearance White Pellets
Assay ≥97.5 % (NaBH4)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR