Sodium borohydride 98+%, powder

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate

AC18930-0050 AC18930-0100 AC18930-1000 AC18930-5000
TS18930-0050EA 61 USD
TS18930-0050 TS18930-0100 TS18930-1000 TS18930-5000
Sodium borohydride 98+%, powder
Sodium borohydride
Formula: NaBH₄
MW: 37.83 g/mol
Melting Pt: 400 (dec.)
Flash Pt: 69
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3 I
Merck Index: 13,08664

Order Now

Specification Test Results

Appearance White Powder
Titration Iodometric ≥98.0 %
Water ≤5 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR