Sodium tetrafluoroborate 97+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium fluoroborate

AC19212-0025 AC19212-0250 AC19212-5000
TS19212-0025EA 282 USD
TS19212-0025 TS19212-0250 TS19212-5000
Sodium tetrafluoroborate 97+%, Extra Pure
Sodium tetrafluoroborate
Formula: BF₄Na
MW: 109.79 g/mol
Melting Pt: 384 °C
Storage Temperature: Ambient
MDL Number: MFCD00003515
CAS Number: 13755-29-8
EINECS: 237-340-6
UN: 1759
ADR: 8,II
Merck Index: 12,08761

Order Now

Specification Test Results

Appearance White to light gray Crystalline powder or crystals
Titration ≥97.0 %
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤1000 ppm
Iron (Fe) ≤30 ppm
Heavy metals (as Pb) ≤30 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR