Nitrobenzene 99%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

AC12842-0010 AC12842-0025 AC12842-0100
TS12842-0010EA 117 USD
TS12842-0010 TS12842-0025 TS12842-0100
Nitrobenzene 99%, extra pure
Nitrobenzene
Formula: C₆H₅NO₂
MW: 123.11 g/mol
Boiling Pt: 210.8
Melting Pt: 5.7
Density: 1.205 g/cm³
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00007043
CAS Number: 98-95-3
EINECS: 202-716-0
UN: 1662
ADR: 6.1 II
Merck Index: 13,06621

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5500 to 1.5540 (20°C, 589 nm)
Water ≤0.15 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR