Magnesium nitrate hexahydrate 98+% ACS
Supplier: Thermo Scientific Chemicals
AC42388-0025
AC42388-5000
TS42388-0025EA
255
USD
TS42388-0025
TS42388-5000
Magnesium nitrate hexahydrate 98+% ACS
Magnesium nitrate hexahydrate
Formula:
Mg(NO₃)₂·6H₂O MW: 256.41 g/mol Boiling Pt: 330 °C (760 mmHg) Melting Pt: 89 °C Flash Pt: 93.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149779 CAS Number: 13446-18-9 EINECS: 233-826-7 Merck Index: 13,05697 |
Specification Test Results
Appearance | Colorless to white Adhering crystalline powder or crystals |
Titration Complexometric | 98.0 to 102.0 % |
pH | 5.0 to 8.2 (5% soln. at 25°C) |
Chloride (Cl) | ≤0.001 % |
Phosphate (PO4) | ≤5 ppm |
Sulfate (SO4) | ≤0.005 % |
Ammonium (NH4) | ≤0.003 % |
Barium (Ba) | ≤0.005 % |
Calcium (Ca) | ≤0.01 % |
Iron (Fe) | ≤5 ppm |
Manganese (Mn) | ≤5 ppm |
Potassium (K) | ≤0.005 % |
Sodium (Na) | ≤0.005 % |
Strontium (Sr) | ≤0.005 % |
Heavy metals (ICP-OES) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...