Sodium sulfate decahydrate, extra pure
Supplier: Thermo Scientific Chemicals
AC12501-0010
AC12501-0025
AC12501-0250
AC12501-2500
TS12501-0010EA
72
USD
TS12501-0010
TS12501-0025
TS12501-0250
TS12501-2500
Sodium sulfate decahydrate, extra pure
Sodium sulphate decahydrate
Formula:
Na₂SO₄·10H₂O Melting Pt: 884 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149189 CAS Number: 7727-73-3 |
Specification Test Results
Appearance | White Crystalline powder or crystals |
pH | 5.2 to 9.2 (5% solution) |
Titration after Ion exchange | ≥99.0 % |
Chloride (Cl) | ≤0.02 % |
Calcium (Ca) | ≤0.01 % |
Iron (Fe) | ≤0.005 % |
Potassium (K) | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...