UniStore™ Large Reagent Bottles, Narrow Mouth, PP
Supplier: United Scientific Supplies
BNMP2L BNMP2L-PK/6 BNMP4L BNMP4L-PK/6 BNMP8L BNMP8L-PK/6
470332-680EA 21.75 USD
470332-680 470332-682 470332-684 470332-686 470332-688 470332-690
UniStore™ Large Reagent Bottles, Narrow Mouth, PP
Bottles Plastic Bottles
Large size reagent bottles.
  • Autoclavable
UniStore™ narrow mouth PP large capacity reagent bottles feature narrow mouths, integrated shoulder loops (except 2 L). Constructed of polypropylene, the bottles are translucent, pliable, and have excellent strength for extended use. Ideal for packing, shipping, and storage purposes.
Ordering information: Lined HDPE cap for a tight seal included.