SMARTouch™ Microinjection Syringe Pump, World Precision Instruments
Supplier: Avantor
MSPP-UMP3T-1EA 3536.1 USD
MSPP-UMP3T-1
SMARTouch™ Microinjection Syringe Pump, World Precision Instruments
Pumps Syringe Pumps
The UltraMicroPump3 (UMP3) is a versatile microinjection syringe pump which uses micro syringes to deliver picoliter to milliliter volumes. The microinjection syringe pump is optimum for applications that require injections of precise and small amounts of liquid. With its touchscreen controller, UMP3 microinjection syringe pump can displace as little as 0.53 µl/step (using 10 µl syringe with 60 mm scale length).
  • Graphic display with SMARTouch™ touch screen
  • Splash proof touch screen
  • User configurable mounting bar
  • Dual mode motor drive
  • Optional foot switch available
  • 5 digit display
Accepts a wide range of microinjection syringes from 0.5 to 1000 µl. Manual or automated injections. Quiet operation for electrophysiology recordings.
Mounts directly on micromanipulator or stereotaxic frame. Rapid setup with intuitive touchscreen controller.