Screen Quest™ Fluo-4 No Wash Calcium Assay Kit, AAT Bioquest
Supplier: AAT BIOQUEST INC
36325 36326
76484-282EA 500.94 USD
76484-282 76484-284
Screen Quest™ Fluo-4 No Wash Calcium Assay Kit, AAT Bioquest
Assays Cellular Assays
Calcium flux assays are preferred methods in drug discovery for screening G protein coupled receptors (GPCR).
  • Well-cited calcium probe
  • Contains all necessary components
  • No wash step required
  • Robust and adaptable protocol
  • Multiple research applications
Caution: For research use only.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=720781854f5fc74a180dfcaaa8590009cbf5fe3b&vid=c6130d9b51f88d616004b373c84e8d717cc1a860&prid=web&clid=SAPDAM