Anti-IgG Goat Polyclonal Antibody (iFluor® 633)
Supplier: AAT BIOQUEST INC
16478 16743
76482-446EA 96.26 USD
76482-446 76482-448
Anti-IgG Goat Polyclonal Antibody (iFluor® 633)
Antibodies
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins
Caution: For research use only.