Ward's® Live Red Worm Culture (Eisenia Hortensis)
Supplier: DMF BAIT COMPANY
25BRW30 BRW500
470336-804CS 289.95 USD
470336-804 470337-638
Ward's® Live Red Worm Culture (Eisenia Hortensis)
Specimens Live Specimens Animals - Living Material
These common worms are also referred to as Red Wigglers.
  • Great to use in composting and fishing
  • Helps you go green, as they are great for composting
  • Chemical-free; safe for feeding to pets
Also known as euro worms, Belgian worms, Belgian reds, minis, and reds. While they are very easy to keep alive for weeks at room temperature in the cups they arrive in, we recommend if not using immediately to refrigerate between 38 to 55 °F.
Ordering information: For larger orders please give at least 2 weeks notice.
Delivery information: These products contain living materials and will ship via overnight or 2 nd day delivery. Please specify a delivery date during checkout.
Packaging: Each cup of worms come packed with sphagnum peat moss.