Temozolomide
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,4-Dihydro-3-methyl-4-oxoimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxamide, 3-Methyl-4-oxo-3,4-dihydroimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxamide
46676-0010
466760050
TS46676-0010EA
150
USD
TS46676-0010
TS466760050
Temozolomide
Temozolomide
Formula:
C₆H₆N₆O₂ MW: 194.15 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00866492 CAS Number: 85622-93-1 Merck Index: 13.9218 |
Specification Test Results
Appearance | White to light pink to brown powder |
Infrared spectrum | Conforms |
Loss on drying | ≤ 1% |
HPLC | ≥ 97.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...