DAF-2 diacetate for nitric oxide detection

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 2-​(3,​6-​diacetyloxy-​4,​5-​diamino-​9H-​xanthen-​9-​yl)-​benzoic acid, DAF-2 DA, 4,5-Diaminofluorescein diacetate

16293 16294
76485-342EA 237.74 USD
76485-342 76485-344
DAF-2 diacetate for nitric oxide detection
DAF-2 diacetate

DAF-2 diacetate is a more cell-permeable version of DAF-2, a sensitive fluorescent indicator for the detection and bioimaging of nitric oxide (NO).


  • Sensitive NO detection

Formula: C₂₄H₁₈N₂O₇
MW: 446.41 g/mol
MDL Number: MFCD05865101
CAS Number: 205391-02-2
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfd9e28eb531c9992e58c1bdf8ed92c2c5f221e1&vid=d7ccd311dd7e94ba6178a7a48a58cf45d84b932b&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae3f4256803977fe1ae3c9e1190e772ff7ec8daa&vid=ae3f4256803977fe1ae3c9e1190e772ff7ec8daa&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR