Streptavidin, RPE
Supplier: AAT BIOQUEST INC
16900 16901
76482-698EA 117.59 USD
76482-698 76482-700
Streptavidin, RPE
Proteins and Peptides
Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin.
  • Extremely high biotin-binding affinity
  • High quality conjugate
Caution: For research use only.