Bio-Paper Biopsy Wraps, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62326-20 62326-21 62326-22 62326-23
76499-548CS 45.18 USD
76499-548 76499-550 76499-552 76499-554
Bio-Paper Biopsy Wraps, Electron Microscopy Sciences
Biopsy Pads
Biopsy wraps are filter paper that is cut to ready-to-use sizes.
  • Packaged in convenient dispensers
Blue Bio-Paper makes it easier to locate small specimens after processing.