Brady® B33 Series Clear with White Printable Zone Vinyl Labels, Brady
Supplier: Brady Worldwide
B33-114-427 B33-117-427
76520-272RL 161.66 USD
76520-272 76520-274
Brady® B33 Series Clear with White Printable Zone Vinyl Labels, Brady
Labels
Self-Laminating Vinyl (B-427) is an excellent material for wire and cable identification. This material has good clarity, conformability, and is self-extinguishing.
  • Recommended ribbon: B30-R4300
  • Self laminating, wrap around labels
  • Permanent acrylic adhesive type
  • Self-laminating vinyl material
Provided in a self-laminated format with a white printable zone and translucent overlaminating area to protect text. Durable, permanent acrylic adhesive has outstanding water and oil resistance. Service temperature of −40 to 158 °F (−40 to 70 °C). A ribbon is required to print this material.