Orum International, Camera CFU Photo, Hardy Diagnostics
Supplier: Hardy Diagnostics
BAS337
76470-206EA 5428.43 USD
76470-206
Orum International, Camera CFU Photo, Hardy Diagnostics
Cameras
Capture photos of culture media after CFU counts are obtained from active impact air sampling.
Integrates with software 76446-766 to maintain unalterable records of results and validate the accuracy of plate counts.