CP-Sil 5 CB Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CP7684 CP7680 CP7682 CP8741 CP6666 CP6670 CP7135 CP7140 CP7170 CP7475 CP7529 CP7625 CP7640 CP7670 CP7675 CP7685 CP7687 CP7690 CP7692 CP7693 CP7709 CP7710 CP7739 CP7750 CP7760 CP8674 CP8677 CP8688 CP8688I5 CP8690 CP8735 CP8770 CP8780 CP8870
AGCP7684EA 390 USD
AGCP7684 AGCP7680 AGCP7682 AGCP8741 AGCP6666 AGCP6670 AGCP7135 AGCP7140 AGCP7170 AGCP7475 AGCP7529 AGCP7625 AGCP7640 AGCP7670 AGCP7675 AGCP7685 AGCP7687 AGCP7690 AGCP7692 AGCP7693 AGCP7709 AGCP7710 AGCP7739 AGCP7750 AGCP7760 AGCP8674 AGCP8677 AGCP8688 AGCP8688I5 AGCP8690 AGCP8735 AGCP8770 AGCP8780 AGCP8870
CP-Sil 5 CB Columns, Agilent Technologies
Chromatography Columns
A low-polarity, general-purpose, 100% dimethylpolysiloxane (PDMS) column ideal for a wide range of applications.
  • 100% dimethylpolysiloxane (PDMS)
  • Nonpolar
  • General purpose phase
  • Bonded, crosslinked, and solvent rinsible
  • Separates almost entirely based on boiling points, making this column suitable for a wide range of applications with a broad temperature range
  • Available in Fused Silica or UltiMetal
  • Tested against the strictest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: SPB-1, Rtx-1, BP-1, OV-1, OV-101, 007-1(MS), SP-2100, SE-30, ZB-1, AT-1, ZB-1
Agilent J&W CP-Sil 5 CB is a general-purpose, nonpolar column containing a 100% dimethylpolysiloxane (PDMS) phase. Separation is almost entirely based on boiling points, making this column suitable for a wide range of applications with a broad temperature range.
Due to extensive crosslinking, CP-Sil 5 CB is highly inert and withstands large solvent injections, guaranteeing reproducibility and ensuring maximum column lifetime. Available in UltiMetal for the highest operating temperatures.