J&W DB-VRX Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
122-1534 122-1564 123-1534 124-1574 122-1564E 123-1564 124-1534
AG122-1534EA 801 USD
AG122-1534 AG122-1564 AG123-1534 AG124-1574 AG122-1564E AG123-1564 AG124-1534
J&W DB-VRX Columns, Agilent Technologies
Chromatography Columns
Features unique selectivity, engineered for optimum resolution of volatiles.
  • Unique selectivity engineered for optimum resolution of volatiles analysis
  • U.S. EPA Methods 502.2, 524.2 and 8260
  • 0.45 mm int.Ø columns provide more plates per meter compared to 0.53 mm id columns for the fewest co-elutions for GC method
  • Doesn't require subambient cooling to resolve the six "gases"
  • Fast run time: <30 minutes for optimum sample throughput, <8 minutes with 0.18 mm id
  • Low polarity
  • Excellent peak shape
  • Bonded, crosslinked, and solvent rinsable
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-VRX features unique selectivity, engineered for optimum resolution of volatiles. These low-polarity columns deliver excellent peak shapes and are fully compliant with U.S. EPA methods 502.2, 524.2, and 8260. They are bonded and crosslinked, and require no subambient cooling to resolve the six 'gases'. DB-VRX features an operating temperature range of –20 to 260 °C.