Anti-Ly6G Mouse Monoclonal Antibody [clone: 1A8]

Supplier: Bio X Cell
BE0075-1-5MG BE0075-1-50MG BE0075-1-25MG BE0075-1-100MG
103252-494EA 3920 USD
103252-494 103252-496 103252-498 103252-500 76555-212 76555-996 76556-532 76557-338
Anti-Ly6G Mouse Monoclonal Antibody [clone: 1A8]
Antibodies
InVivoMab anti-mouse Ly6G, Clone: RB6-8C5, Isotype: rat IgG2a, κ, Applications: in vivo neutrophil depletion, in vivo MDSC depletion, Immunofluorescence, Immunohistochemistry (paraffin), Immunohistochemistry (frozen), Flow cytometry

Type: Primary
Antigen: Ly6G
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR