Matrine
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(7aS,13aR,13bR,13cS)-Dodecahydro-1H,5H,10H-dipyrido[2,1-f:3',2',1'-ij][1,6]naphthyridin-10-one, Matridin-15-one
46659-2500
466590010
TS46659-2500EA
115
USD
TS46659-2500
TS466590010
Matrine
Matrine
Formula:
C₁₅H₂₄N₂O MW: 248.37 g/mol Melting Pt: 76°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00210527 CAS Number: 519-02-8 |
Specification Test Results
Appearance | White to yellow powder |
Infrared spectrum | Conforms |
Water | ≤ 5% |
GC | ≥ 97.0% |
Specific optical rotation | +31.0° to +36.0° (20°C, 589nm) (c=1, EtOH, dry substance) |
Solubility | (5% water) Clear colourless to pale yellow solution |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...