Tetramethylammonium hydroxide 0.26 N semiconductor grade NOVO DEVELOPER 26
Supplier: Transene
|
Danger
|
834-GAL
834-QT
76540-994CS
175.44
USD
76540-994
76541-054
Tetramethylammonium hydroxide 0.26 N semiconductor grade NOVO DEVELOPER 26
Tetramethylammonium hydroxide
Transene NOVO Developers are available in standard concentrations for controlled developing of photoresists.
Formula:
C₄H₁₃NO MW: 91.15 Boiling Pt: 102 °C Melting Pt: 0 °C Density: 1.01 Storage Temperature: Ambient |
MDL Number:
MFCD00008280 CAS Number: 75-59-2 UN: 1835 ADR: 8 II Merck Index: 14,09224 |
Specification Test Results
TMAH Concentration | 2.27-2.47 % |
TMAH Normality | 0,26 Nominal |
Typical Development Cycle | |
Developer | 15 to 75 Seconds |
Water Rinse | 20 to 30 Seconds |
Nitrogen Dry | 15 to 20 Seconds |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...