D-(-)-Luciferin ≥95%
Supplier: AAT BIOQUEST INC
|
Warning
|
12501
12502
12503
76481-988EA
96.26
USD
76481-988
76481-990
76481-992
D-(-)-Luciferin ≥95%
D(-)-Luciferin
Luciferin is the most popular and versatile bioluminescent substrate.
Formula:
C₁₁H₈N₂O₃S₂ |
MDL Number:
MFCD00042929 CAS Number: 2591-17-5 ADR: 5,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...