Tetramethylrhodamine 6-isothiocyanate chloride ≥90%

Supplier: AAT Bioquest
Warning

417 418
76480-888EA 249.58 USD
76480-888 76480-890
Tetramethylrhodamine 6-isothiocyanate chloride ≥90%
Tetramethylrhodamine 6-isothiocyanate chloride (6-TRITC)
Formula: C₂₅H₂₂ClN₃O₃S
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c25a9ea56b575f59350a2a368f8779c4e033485&vid=b9dbf62776c36a6d4faf443deb24fdc871275cb6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR