5(6)-Tetramethylrhodamine isothiocyanate [5(6)-TRITC] ≥90%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: Tetramethylrhodamine-5(6)-isothiocyanate, 5(6)-TRITC, MRITC, TRITC

409 410
76480-878EA 500.94 USD
76480-878 76480-880
5(6)-Tetramethylrhodamine isothiocyanate [5(6)-TRITC] ≥90%
5(6)-Tetramethylrhodamine isothiocyanate [5(6)-TRITC]

5(6)-TRITC is an amino-reactive labeling reagent that is widely used in preparing bioconjugates of proteins and nucleic acids.

Formula: C₂₅H₂₁N₃O₃S
MDL Number: MFCD00063517
CAS Number: 95197-95-8
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a4608521e6bb3151793af30309e044bdac4417c&vid=d5a171979e5a3dbb9bbf12627ab5f5f10f77a5c1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR