5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE] ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: NHS-Rhodamine, NHS-Rhoda, TAMRA SE, 5 and 6 isomers, TMR-SE, 5(6)-Carboxytetramethylrhodamine N-succinimidyl ester, 5(6)-TAMRA N-succinimidyl ester

370 371 372
76480-836EA 124.31 USD
76480-836 76480-838 76480-840
5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE] ≥95%
5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE]

The succinimidyl esters of 5-TAMRA, 6-TAMRA or the mixed isomers are the primary labeling reagents for the preparation of orange fluorescent bioconjugates, including peptide, protein, nucleotide and nucleic acid conjugates, especially fluorescent antibodies and avidin derivatives used in immunochemistry.

Formula: C₂₉H₂₅N₃O₇
MW: 527.52 g/mol
MDL Number: MFCD00077324
CAS Number: 150408-83-6
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2170a7b242e001cec4beadef49801e7c36812f37&vid=e59b08f03e44ff31fbab8cb46d13c7c8a2d20e40&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR