5(6)-TAMRA ≥95%

Supplier: AAT Bioquest
Warning

360 361 362
76480-818EA 124.31 USD
76480-818 76480-820 76480-822
5(6)-TAMRA ≥95%
5(6)-TAMRA
Formula: C₂₅H₂₂N₂O₅
MDL Number: MFCD16661194
CAS Number: 98181-63-6
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a59cacafc8cb51210444c76e4e24af3b67c0c4c&vid=aa59b976796fe44f82fd97dd98afd367c3601b6e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR