6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE) ≥95%

Supplier: AAT Bioquest
Warning

116 117 118
76480-572EA 124.31 USD
76480-572 76480-574 76480-576
6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE) ≥95%
6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE)

6-FAM, SE is the amine-reactive succinimidyl ester of single isomer 6-FAM acid.

Formula: C₂₅H₁₅NO₉
MDL Number: MFCD00077323
CAS Number: 92557-81-8
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=075c29e63190b5c01b15c0d616d98ecf6fe5cdf9&vid=dc1eacf64c004320f7c83449b710c3e253a1873e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR