6-Carboxyfluorescein ≥95%
Supplier: AAT BIOQUEST INC
|
Warning
|
Synonyms:
3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-6-carboxylic acid, 6-FAM
106
107
108
76480-554EA
96.26
USD
76480-554
76480-556
76480-558
6-Carboxyfluorescein ≥95%
6-Carboxyfluorescein
6-FAM is the other purified isomer of 5(6)-carboxyfluorescein.
Formula:
C₂₁H₁₂O₇ MW: 376.32 g/mol |
MDL Number:
MFCD00036873 CAS Number: 3301-79-9 ADR: 5 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...