Cysteaminium chloride, Pierce™

Supplier: 0000013509

Synonyms: Cysteamine HydrochlorideMercaptoethylamine-HClβ-Mercaptoethylamine hydrochlorideH-Cys-NH2·HCl2-Aminoethanethiol hydrochloride2-MEA2-Mercaptoethylamine hydrochloride

20408 20408
PI20408EA 293.69 USD
CAPI20408 PI20408
Cysteaminium chloride, Pierce™
Cysteaminium chloride

Thermo Scientific Pierce 2-Mercaptoethylamine-HCl, commonly abbreviated 2-MEA, is a mild reductant that is often used to selectively reduce hinge-region disulfide bonds in antibody.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Formula: C₂H₇NS·HCl
MW: 113.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00012904
CAS Number: 156-57-0
EINECS: 205-858-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR