iFluor® 594, Succinimidyl Ester
Supplier: AAT BIOQUEST INC
1029 71029 71508 71558
76481-090EA 249.58 USD
76481-090 76481-092 76481-094 76481-096
iFluor® 594, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Caution: For Research Use Only