Brownlee Electrolysis Apparatus (with Beaker)
Supplier: United Scientific Supplies
BEA001-B BEA001
470325-642CS 34.95 USD
470325-642 470339-714
Brownlee Electrolysis Apparatus (with Beaker)
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Small electrolysis apparatus with beaker included.
  • Simple and economical electrolysis apparatus
  • Contains platinum electrodes, insulated connecting rods, binding posts, and a nonconducting support
  • Beaker has two clips to hold an inverted test tube
Operates on a user-supplied 6 V battery or 10 V DC power supply.